Assay Details
| Gene Symbol: Enox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271447.1 | NP_001258376.1 | 3 - 4 | 344 | 114 | |
| NM_001271449.1 | NP_001258378.1 | 3 - 4 | 333 | 114 | ||
| NM_001271450.1 | NP_001258379.1 | 3 - 4 | 333 | 114 | ||
| NM_001271451.1 | NP_001258380.1 | 3 - 4 | 333 | 114 | ||
| NM_145951.5 | NP_666063.1 | 3 - 4 | 288 | 114 | ||
| GenBank mRNA | AK028343.1 | - | 3 - 4 | 334 | 114 | |
| AK034058.1 | - | 3 - 4 | 289 | 114 | ||
| AK041822.1 | - | 3 - 4 | 334 | 114 | ||
| AK077508.1 | - | 3 - 4 | 315 | 114 | ||
| AK137736.1 | - | 3 - 4 | 315 | 114 | ||
| AK139897.1 | - | 3 - 4 | 315 | 114 | ||
| BC025915.1 | - | 3 - 4 | 307 | 5043650 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034058.1, AK041822.1, AK137736.1, AK139897.1, BC025915.1, NM_001271447.1, NM_001271449.1, NM_001271450.1, NM_001271451.1, NM_145951.5 transcripts.
Set Membership: |
|

Enox2
Genomic Map