Assay Details
| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178775.4 | NP_848890.3 | 3 - 4 | 375 | 81 | |
| GenBank mRNA | AK030313.1 | - | 353 | 81 | ||
| AK149858.1 | - | 3 - 4 | 340 | 81 | ||
| BB619353.1 | - | 3 - 4 | 376 | 81 | ||
| CJ045347.1 | - | 3 - 4 | 376 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kc1
Genomic Map