Assay Details
| Gene Symbol: Kctd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017384.1 | - | 2 - 3 | 266 | 86 | |
| AK082924.1 | - | 2 - 3 | 265 | 86 | ||
| AK159067.1 | - | 2 - 3 | 1678 | 86 | ||
| BB625950.1 | - | 2 - 3 | 155 | 86 | ||
| BC096370.1 | - | 2 - 3 | 105 | 3597940 | 86 | |
| BC131971.1 | - | 1 - 2 | 75 | 40130248 | 86 | |
| BC131975.1 | - | 1 - 2 | 75 | 40130252 | 86 | |
| CX239507.1 | - | 2 - 3 | 332 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK082924.1,AK159067.1,BC096370.1,BC131971.1,BC131975.1 transcripts.
This assay is in the 5'UTR region of the AK082924.1,AK159067.1,BC096370.1,BC131971.1,BC131975.1 transcripts.
Set Membership: |
|

Kctd6
Genomic Map