Assay Details
| Gene Symbol: Hhat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144881.4 | NP_659130.2 | 5 - 6 | 750 | 71 | |
| GenBank mRNA | AK028230.1 | - | 5 - 6 | 553 | 71 | |
| AK134661.1 | - | 5 - 6 | 593 | 71 | ||
| AK172126.1 | - | 5 - 6 | 597 | 71 | ||
| BC008159.1 | - | 744 | 3963716 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hhat
Genomic Map