Assay Details
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093752.1 | NP_001087221.1 | 3 - 4 | 453 | 75 | |
| NM_001093753.1 | NP_001087222.1 | 2 - 3 | 435 | 75 | ||
| GenBank mRNA | AK011472.1 | - | 3 - 4 | 391 | 75 | |
| AK029394.1 | - | 2 - 3 | 398 | 75 | ||
| AK146719.1 | - | 2 - 3 | 407 | 75 | ||
| AK183987.1 | - | 1 - 2 | 136 | 75 | ||
| AK218039.1 | - | 1 - 2 | 136 | 75 | ||
| BC002111.1 | - | 2 - 3 | 406 | 75 | ||
| BC021809.1 | - | 2 - 3 | 390 | 75 | ||
| BC037053.1 | - | 3 - 4 | 376 | 75 | ||
| BC038928.1 | - | 3 - 4 | 412 | 75 | ||
| BC055763.1 | - | 4 - 5 | 341 | 75 | ||
| BC069945.1 | - | 2 - 3 | 385 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srsf11
Genomic Map