Assay Details
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093752.1 | NP_001087221.1 | 7 - 8 | 830 | 88 | |
| NM_001093753.1 | NP_001087222.1 | 6 - 7 | 812 | 88 | ||
| GenBank mRNA | AK011472.1 | - | 7 - 8 | 768 | 88 | |
| AK029394.1 | - | 6 - 7 | 775 | 88 | ||
| AK081511.1 | - | 4 - 5 | 637 | 88 | ||
| AK146719.1 | - | 6 - 7 | 784 | 88 | ||
| BC002111.1 | - | 6 - 7 | 783 | 88 | ||
| BC021809.1 | - | 6 - 7 | 767 | 88 | ||
| BC037053.1 | - | 7 - 8 | 753 | 88 | ||
| BC038928.1 | - | 7 - 8 | 789 | 88 | ||
| BC055763.1 | - | 8 - 9 | 87 | |||
| BC069945.1 | - | 6 - 7 | 762 | 88 | ||
| BC138145.1 | - | 4 - 5 | 565 | 8861164 | 88 | |
| BC145162.1 | - | 4 - 5 | 565 | 9053713 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srsf11
Genomic Map