Assay Details
Gene Symbol: Lrrc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289524.1 | NP_001276453.1 | 13 - 14 | 1561 | 98 | |
NM_001289525.1 | NP_001276454.1 | 11 - 12 | 1308 | 98 | ||
NM_024194.6 | NP_077156.2 | 13 - 14 | 1564 | 98 | ||
GenBank mRNA | AK011726.1 | - | 13 - 14 | 1545 | 98 | |
AK017814.1 | - | 13 - 14 | 1533 | 98 | ||
AK034978.1 | - | 1544 | 98 | |||
AK148748.1 | - | 13 - 14 | 1524 | 98 | ||
AK152244.1 | - | 11 - 12 | 1289 | 98 | ||
AK163288.1 | - | 13 - 14 | 1532 | 98 | ||
AK170052.1 | - | 13 - 14 | 1541 | 98 | ||
BC002279.1 | - | 9 - 10 | 953 | 3497400 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
