Assay Details
| Gene Symbol: Pde10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290707.1 | NP_001277636.1 | 17 - 18 | 2031 | 90 | |
| NM_011866.2 | NP_035996.2 | 16 - 17 | 1985 | 90 | ||
| GenBank mRNA | AF110507.1 | - | 16 - 17 | 1856 | 90 | |
| AK039249.1 | - | 17 - 18 | 2031 | 90 | ||
| AK147690.1 | - | 15 - 16 | 1902 | 90 | ||
| AK162221.1 | - | 16 - 17 | 1640 | 90 | ||
| AK162804.1 | - | 14 - 15 | 1526 | 90 | ||
| AK166310.1 | - | 8 - 9 | 997 | 90 | ||
| AY360383.1 | - | 16 - 17 | 1985 | 90 | ||
| BC113201.1 | - | 16 - 17 | 1851 | 40048238 | 90 | |
More Information
Set Membership: |
|

Pde10a
Genomic Map