Assay Details
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008246.2 | NP_032272.2 | 10 - 11 | 1300 | 86 | |
GenBank mRNA | AK004779.1 | - | 10 - 11 | 1239 | 86 | |
AK135865.1 | - | 10 - 11 | 1121 | 86 | ||
AK158838.1 | - | 10 - 11 | 1301 | 86 | ||
AK169626.1 | - | 10 - 11 | 1240 | 86 | ||
BC118047.1 | - | 10 - 11 | 1120 | 40106779 | 86 | |
D88315.1 | - | 10 - 11 | 1173 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
