Assay Details
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008246.2 | NP_032272.2 | 3 - 4 | 438 | 119 | |
GenBank mRNA | AK004779.1 | - | 3 - 4 | 377 | 119 | |
AK135865.1 | - | 3 - 4 | 259 | 119 | ||
AK158838.1 | - | 3 - 4 | 439 | 119 | ||
AK169626.1 | - | 3 - 4 | 378 | 119 | ||
BC118047.1 | - | 3 - 4 | 258 | 40106779 | 119 | |
D88315.1 | - | 3 - 4 | 311 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
