Assay Details
| Gene Symbol: Frrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113478.1 | NP_001106950.1 | 5 - 6 | 607 | 76 | |
| NM_009146.3 | NP_033172.2 | 5 - 6 | 569 | 76 | ||
| GenBank mRNA | AK075573.1 | - | 5 - 6 | 561 | 76 | |
| AK144933.1 | - | 5 - 6 | 561 | 76 | ||
| AK162693.1 | - | 5 - 6 | 562 | 76 | ||
| AK167289.1 | - | 5 - 6 | 597 | 76 | ||
| BC027770.1 | - | 4 - 5 | 567 | 5344922 | 76 | |
| D50464.1 | - | 5 - 6 | 557 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frrs1
Genomic Map