Assay Details
| Gene Symbol: Ttll7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014905.1 | - | 5 - 6 | 710 | 98 | |
| AM690750.1 | - | 4 - 5 | 351 | 98 | ||
| AM690751.1 | - | 4 - 5 | 351 | 98 | ||
More Information
Set Membership: |
|

Ttll7
Genomic Map