Assay Details
| Gene Symbol: Apba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291166.1 | NP_001278095.1 | 6 - 7 | 1405 | 92 | |
| NM_001291167.1 | NP_001278096.1 | 5 - 6 | 1351 | 92 | ||
| NM_007461.2 | NP_031487.1 | 6 - 7 | 1402 | 92 | ||
| GenBank mRNA | BC057620.1 | - | 6 - 7 | 1402 | 6402377 | 92 |
| BC060269.1 | - | 3 - 4 | 1105 | 6810117 | 92 | |
| L34676.1 | - | 3 - 4 | 864 | 92 | ||
More Information
Set Membership: |
|

Apba2
Genomic Map