Assay Details
| Gene Symbol: Apba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291166.1 | NP_001278095.1 | 10 - 11 | 1866 | 64 | |
| NM_001291167.1 | NP_001278096.1 | 9 - 10 | 1812 | 64 | ||
| NM_007461.2 | NP_031487.1 | 10 - 11 | 1863 | 64 | ||
| GenBank mRNA | BC057620.1 | - | 10 - 11 | 1863 | 6402377 | 64 |
| BC060269.1 | - | 7 - 8 | 1566 | 6810117 | 64 | |
| L34676.1 | - | 7 - 8 | 1325 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apba2
Genomic Map