Assay Details
| Gene Symbol: Apba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291166.1 | NP_001278095.1 | 11 - 12 | 2041 | 75 | |
| NM_001291167.1 | NP_001278096.1 | 10 - 11 | 1987 | 75 | ||
| NM_007461.2 | NP_031487.1 | 11 - 12 | 2038 | 75 | ||
| GenBank mRNA | BC057620.1 | - | 11 - 12 | 2038 | 6402377 | 75 |
| BC060269.1 | - | 8 - 9 | 1741 | 6810117 | 75 | |
| L34676.1 | - | 8 - 9 | 1500 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apba2
Genomic Map