Assay Details
| Gene Symbol: Apba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291166.1 | NP_001278095.1 | 12 - 13 | 2252 | 62 | |
| NM_001291167.1 | NP_001278096.1 | 11 - 12 | 2198 | 62 | ||
| NM_007461.2 | NP_031487.1 | 12 - 13 | 2249 | 62 | ||
| GenBank mRNA | AK158629.1 | - | 2 - 3 | 339 | 62 | |
| BC057620.1 | - | 12 - 13 | 2249 | 6402377 | 62 | |
| BC060269.1 | - | 9 - 10 | 1952 | 6810117 | 62 | |
| L34676.1 | - | 9 - 10 | 1711 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apba2
Genomic Map