Assay Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172742.2 | NP_766330.2 | 3 - 4 | 366 | 137 | |
| GenBank mRNA | AK088715.1 | - | 3 - 4 | 384 | 137 | |
| BC041672.1 | - | 3 - 4 | 338 | 137 | ||
| CK634659.1 | - | 3 - 4 | 252 | 137 | ||
| CN669724.1 | - | 3 - 4 | 366 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr10
Genomic Map