Assay Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172742.2 | NP_766330.2 | 6 - 7 | 679 | 87 | |
| GenBank mRNA | AK214447.1 | - | 2 - 3 | 115 | 87 | |
| BC041672.1 | - | 6 - 7 | 651 | 87 | ||
| BC055074.1 | - | 4 - 5 | 343 | 30052208 | 87 | |
| CF734341.1 | - | 1 - 2 | 58 | 88 | ||
| CK634659.1 | - | 6 - 7 | 565 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr10
Genomic Map