Assay Details
| Gene Symbol: Adgrl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133222.3 | NP_573485.2 | 6 - 7 | 818 | 104 | |
| GenBank mRNA | AF385682.1 | - | 6 - 7 | 821 | 104 | |
| BC017134.1 | - | 6 - 7 | 756 | 4511572 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgrl4
Genomic Map