Assay Details
| Gene Symbol: Uox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009474.5 | NP_033500.1 | 5 - 6 | 687 | 63 | |
| GenBank mRNA | AK050047.1 | - | 4 - 5 | 477 | 63 | |
| AK050425.1 | - | 5 - 6 | 688 | 63 | ||
| BC019771.1 | - | 5 - 6 | 669 | 5136328 | 63 | |
| M27695.1 | - | 5 - 6 | 642 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uox
Genomic Map