Assay Details
| Gene Symbol: Tmc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177695.4 | NP_808363.3 | 4 - 5 | 560 | 81 | |
| GenBank mRNA | AY236491.1 | - | 4 - 5 | 564 | 81 | |
| AY263157.1 | - | 4 - 5 | 561 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmc3
Genomic Map