Assay Details
Gene Symbol: Mcoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005846.2 | NP_001005846.1 | 6 - 7 | 820 | 95 | |
NM_026656.4 | NP_080932.2 | 6 - 7 | 848 | 95 | ||
GenBank mRNA | AF503575.1 | - | 6 - 7 | 837 | 95 | |
AK014467.1 | - | 6 - 7 | 751 | 95 | ||
AK019454.1 | - | 6 - 7 | 717 | 95 | ||
AK150446.1 | - | 6 - 7 | 816 | 95 | ||
AK150547.1 | - | 6 - 7 | 816 | 95 | ||
AK152067.1 | - | 6 - 7 | 816 | 95 | ||
AK152943.1 | - | 6 - 7 | 816 | 95 | ||
AK153290.1 | - | 6 - 7 | 816 | 95 | ||
AK159172.1 | - | 6 - 7 | 848 | 95 | ||
AY083532.1 | - | 5 - 6 | 666 | 95 | ||
BC029847.1 | - | 6 - 7 | 697 | 3967456 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
