Assay Details
| Gene Symbol: Rap1gds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040690.2 | NP_001035780.1 | 4 - 5 | 528 | 77 | |
| NM_001286759.1 | NP_001273688.1 | 4 - 5 | 528 | 77 | ||
| GenBank mRNA | AK154852.1 | - | 4 - 5 | 514 | 77 | |
| AK155793.1 | - | 4 - 5 | 514 | 77 | ||
| AK160989.1 | - | 4 - 5 | 464 | 77 | ||
| AK164159.1 | - | 4 - 5 | 440 | 77 | ||
| AK166315.1 | - | 4 - 5 | 461 | 77 | ||
| AK172052.1 | - | 4 - 5 | 530 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rap1gds1
Genomic Map