Assay Details
Gene Symbol: Mas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008552.4 | NP_032578.2 | 1 - 2 | 48 | 150 | |
GenBank mRNA | AK015036.1 | - | 1 - 2 | 39 | 150 | |
AK030261.1 | - | 1 - 2 | 38 | 150 | ||
AK030265.1 | - | 1 - 2 | 49 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK015036.1, AK030261.1, AK030265.1, NM_008552.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
