Assay Details
Gene Symbol: Tdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021480.5 | NP_067455.5 | 4 - 5 | 702 | 69 | |
GenBank mRNA | AF134346.2 | - | 4 - 5 | 366 | 69 | |
AK077571.1 | - | 4 - 5 | 377 | 69 | ||
AY116662.1 | - | 3 - 4 | 303 | 69 | ||
BC058860.1 | - | 4 - 5 | 717 | 30014660 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
