Assay Details
| Gene Symbol: Pinx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028228.3 | NP_082504.1 | 3 - 4 | 337 | 63 | |
| GenBank mRNA | AF321817.1 | - | 3 - 4 | 285 | 63 | |
| AF421879.1 | - | 3 - 4 | 220 | 63 | ||
| AJ344106.1 | - | 2 - 3 | 109 | 63 | ||
| AK012057.1 | - | 3 - 4 | 315 | 63 | ||
| AK075840.1 | - | 1 - 2 | 107 | 63 | ||
| BC065781.1 | - | 3 - 4 | 236 | 63 | ||
| BC115636.1 | - | 3 - 4 | 249 | 40102952 | 63 | |
| BC115637.1 | - | 3 - 4 | 247 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pinx1
Genomic Map