Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00013740.mm.1 |
NM_001166648.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013740.mm.1 |
NM_001166648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013740.mm.1 |
NM_001166649.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013740.mm.1 |
NM_001166649.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013740.mm.1 |
NM_001166650.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013740.mm.1 |
NM_001166650.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013740.mm.1 |
NM_153532.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013740.mm.1 |
NM_153532.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013740.mm.1 |
TC0X00002166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013743.mm.1 |
NM_153532.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013743.mm.1 |
NM_153532.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013743.mm.1 |
TC0X00002166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013741.mm.1 |
NM_001166648.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013741.mm.1 |
NM_001166648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013741.mm.1 |
NM_001166649.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013741.mm.1 |
NM_001166649.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013741.mm.1 |
NM_001166650.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013741.mm.1 |
NM_001166650.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013741.mm.1 |
NM_153532.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013741.mm.1 |
NM_153532.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013741.mm.1 |
TC0X00002166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013745.mm.1 |
NM_001166648.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013745.mm.1 |
NM_001166648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013745.mm.1 |
TC0X00002166.mm.1 |
MTA TC |