Assay Details
Gene Symbol: Slc9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178877.6 | NP_849208.4 | 10 - 11 | 1389 | 104 | |
GenBank mRNA | AK029417.1 | - | 7 - 8 | 940 | 104 | |
AK039655.1 | - | 1 - 2 | 388 | 104 | ||
BC090977.1 | - | 10 - 11 | 1342 | 30503721 | 104 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK039655.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
