Assay Details
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013667.2 | NP_038695.1 | 4 - 5 | 937 | 138 | |
GenBank mRNA | AJ006036.1 | - | 4 - 5 | 937 | 138 | |
AK052658.1 | - | 4 - 5 | 934 | 138 | ||
AK143892.1 | - | 4 - 5 | 938 | 138 | ||
BC015250.1 | - | 4 - 5 | 865 | 4242038 | 138 | |
BC069911.1 | - | 4 - 5 | 884 | 4974824 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
