Assay Details
| Gene Symbol: Mrpl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026310.3 | NP_080586.1 | 3 - 4 | 564 | 91 | |
| GenBank mRNA | AK003104.1 | - | 3 - 4 | 569 | 91 | |
| AK008680.1 | - | 3 - 4 | 565 | 91 | ||
| AK010150.1 | - | 3 - 4 | 617 | 91 | ||
| AK089039.1 | - | 2 - 3 | 273 | 91 | ||
| BC011425.1 | - | 3 - 4 | 539 | 3603233 | 91 | |
More Information
Set Membership: |
|

Mrpl18
Genomic Map