Assay Details
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025364.3 | NP_001020535.1 | 3 - 4 | 458 | 63 | |
NM_013648.6 | NP_038676.1 | 6 - 7 | 1372 | 63 | ||
GenBank mRNA | AF038538.1 | - | 6 - 7 | 1328 | 63 | |
AK144271.1 | - | 6 - 7 | 1335 | 63 | ||
AK160500.1 | - | 3 - 4 | 419 | 63 | ||
BC031370.1 | - | 6 - 7 | 1329 | 5359358 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
