Assay Details
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025364.3 | NP_001020535.1 | 4 - 5 | 532 | 94 | |
NM_013648.6 | NP_038676.1 | 7 - 8 | 1446 | 94 | ||
GenBank mRNA | AF038538.1 | - | 7 - 8 | 1402 | 94 | |
AF038539.1 | - | 4 - 5 | 471 | 94 | ||
AK144271.1 | - | 7 - 8 | 1409 | 94 | ||
AK160500.1 | - | 4 - 5 | 493 | 94 | ||
BC031370.1 | - | 7 - 8 | 1403 | 5359358 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
