Assay Details
| Gene Symbol: Rccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173445.4 | NP_775621.2 | 6 - 7 | 1704 | 87 | |
| GenBank mRNA | AK030872.1 | - | 6 - 7 | 1675 | 87 | |
| AK047578.1 | - | 849 | 87 | |||
| AK084219.1 | - | 4 - 5 | 1630 | 87 | ||
| AK088662.1 | - | 6 - 7 | 1345 | 87 | ||
| BC069844.1 | - | 6 - 7 | 1684 | 6487223 | 87 | |
More Information
Set Membership: |
|

Rccd1
Genomic Map