| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700030331.mm.1 |
NM_173445.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030331.mm.1 |
NM_173445.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030331.mm.1 |
TC0700003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030336.mm.1 |
NM_173445.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030336.mm.1 |
NM_173445.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030336.mm.1 |
TC0700003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030333.mm.1 |
NM_173445.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030333.mm.1 |
NM_173445.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030333.mm.1 |
TC0700003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030321.mm.1 |
TC0700003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030330.mm.1 |
NM_173445.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030330.mm.1 |
NM_173445.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030330.mm.1 |
TC0700003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030326.mm.1 |
NM_173445.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030326.mm.1 |
NM_173445.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030326.mm.1 |
TC0700003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030327.mm.1 |
NM_173445.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030327.mm.1 |
NM_173445.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030327.mm.1 |
TC0700003590.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030332.mm.1 |
NM_173445.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030332.mm.1 |
NM_173445.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030332.mm.1 |
TC0700003590.mm.1 |
MTA TC |