Assay Details
| Gene Symbol: Myh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164171.1 | NP_001157643.1 | 10 - 11 | 1097 | 74 | |
| NM_010856.4 | NP_034986.1 | 10 - 11 | 1033 | 74 | ||
| GenBank mRNA | AK142232.1 | - | 10 - 11 | 1070 | 74 | |
| BC061145.1 | - | 10 - 11 | 1066 | 74 | ||
| BC090979.1 | - | 10 - 11 | 1066 | 74 | ||
| BC110700.1 | - | 10 - 11 | 1079 | 6786621 | 74 | |
| M76600.1 | - | 8 - 9 | 895 | 74 | ||
| M76601.1 | - | 10 - 11 | 997 | 74 | ||
More Information
Set Membership: |
|

Myh6
Genomic Map