Assay Details
| Gene Symbol: Cdh24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199470.2 | NP_955764.1 | 11 - 12 | 2112 | 65 | |
| GenBank mRNA | BC057373.1 | - | 2066 | 6817232 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613385 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613441 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613382 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613381 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613380 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613442 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613440 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613383 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613438 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061834 | BC057373.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061834 | NM_199470.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061834 | AK006648.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234654 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 253406 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 711676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 21467 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102813 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128013 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061834 | BC057373.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061834 | NM_199470.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061834 | AK006648.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29613431 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128013 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08116323 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102813 | - | - |
| Whole Mouse Genome G4122A | A_52_P487573 | BC057373.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P487573 | NM_199470.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116825 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135862 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102813 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128013 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11677117 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11677122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11677112 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613441 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613438 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613440 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613383 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613442 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613380 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613382 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613385 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29613381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29613388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11134148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11299344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11299341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11529892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11181207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11299338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11134139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11614807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11134152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11246859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11589053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11246858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11134138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29613417 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10419817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419815 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419820 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419824 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419823 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419814 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415090 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400015511.mm.1 | TC1400002062.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015513.mm.1 | NM_199470.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015513.mm.1 | NM_199470.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015513.mm.1 | TC1400002062.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 133508_PM_at | - | - |
| Murine Genome U74v2 Set | 133508_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 700958 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6980592 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006980592 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2699800 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2699800 | - | - |
| mouse-6 v1.1 (Illumina) | scl0239096.2_133-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6980592 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0239096.2_133-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980592 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2699800 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2699800 | - | - |

Cdh24
Genomic Map