Assay Details
| Gene Symbol: Cdh24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199470.2 | NP_955764.1 | 10 - 11 | 1860 | 115 | |
| GenBank mRNA | BC057373.1 | - | 1814 | 6817232 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh24
Genomic Map