Assay Details
| Gene Symbol: Camk1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144817.2 | NP_659066.1 | 11 - 12 | 1485 | 68 | |
| GenBank mRNA | AF428262.1 | - | 11 - 12 | 1433 | 68 | |
| AY212936.1 | - | 10 - 11 | 1365 | 68 | ||
| BC021840.1 | - | 11 - 12 | 1434 | 4502479 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camk1g
Genomic Map