Assay Details
| Gene Symbol: Traf3ip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153137.4 | NP_694777.3 | 10 - 11 | 1338 | 56 | |
| GenBank mRNA | AK031400.1 | - | 10 - 11 | 1278 | 56 | |
| AK161211.1 | - | 7 - 8 | 948 | 56 | ||
| AY383616.1 | - | 8 - 9 | 922 | 56 | ||
| BC036561.1 | - | 10 - 11 | 1146 | 56 | ||
| BC137605.1 | - | 8 - 9 | 949 | 8860624 | 56 | |
| BC171933.1 | - | 8 - 9 | 949 | 9054217 | 56 | |
More Information
Set Membership: |
|

Traf3ip3
Genomic Map