Assay Details
| Gene Symbol: Traf3ip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153137.4 | NP_694777.3 | 7 - 8 | 1020 | 113 | |
| GenBank mRNA | AK031400.1 | - | 7 - 8 | 960 | 113 | |
More Information
Set Membership: |
|

Traf3ip3
Genomic Map