Assay Details
| Gene Symbol: Dusp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022019.5 | NP_071302.2 | 1 - 2 | 103 | 73 | |
| GenBank mRNA | AB037908.1 | - | 1 - 2 | 109 | 73 | |
| AK011995.1 | - | 1 - 2 | 87 | 73 | ||
| AK035293.1 | - | 1 - 2 | 101 | 73 | ||
| AK050528.1 | - | 1 - 2 | 101 | 73 | ||
| AK088024.1 | - | 1 - 2 | 102 | 73 | ||
| AK088357.1 | - | 1 - 2 | 99 | 73 | ||
| AK142568.1 | - | 1 - 2 | 105 | 73 | ||
| BC025066.1 | - | 1 - 2 | 118 | 3495358 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB037908.1, AK011995.1, AK035293.1, AK050528.1, AK088024.1, AK088357.1, AK142568.1, BC025066.1, NM_022019.5 transcripts.
Set Membership: |
|

Dusp10
Genomic Map