Assay Details
| Gene Symbol: Disp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278218.1 | NP_001265147.1 | 4 - 5 | 882 | 67 | |
| NM_001278219.1 | NP_001265148.1 | 4 - 5 | 842 | 67 | ||
| NM_001278220.1 | NP_001265149.1 | 3 - 4 | 725 | 67 | ||
| NM_026866.3 | NP_081142.3 | 5 - 6 | 970 | 67 | ||
| GenBank mRNA | AK004521.1 | - | 3 - 4 | 700 | 67 | |
| AK138276.1 | - | 5 - 6 | 927 | 67 | ||
| AK170113.1 | - | 4 - 5 | 837 | 67 | ||
| AY144589.1 | - | 3 - 4 | 680 | 67 | ||
| BC043102.1 | - | 4 - 5 | 843 | 6406376 | 67 | |
| BC059225.1 | - | 4 - 5 | 843 | 6406376 | 67 | |
More Information
Set Membership: |
|

Disp1
Genomic Map