Assay Details
Gene Symbol: Ndrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145959.1 | NP_001139431.1 | 5 - 6 | 532 | 71 | |
NM_013864.2 | NP_038892.1 | 6 - 7 | 574 | 71 | ||
GenBank mRNA | AB033921.1 | - | 6 - 7 | 437 | 71 | |
AK076514.1 | - | 3 - 4 | 208 | 71 | ||
AK154597.1 | - | 6 - 7 | 442 | 71 | ||
AK158900.1 | - | 5 - 6 | 441 | 71 | ||
AK168011.1 | - | 5 - 6 | 399 | 71 | ||
AK173136.1 | - | 3 - 4 | 215 | 71 | ||
BC012963.1 | - | 6 - 7 | 441 | 4211421 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
