Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400017382.mm.1 |
NM_021458.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017382.mm.1 |
NM_021458.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017382.mm.1 |
TC1400002252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017384.mm.1 |
NM_021458.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017384.mm.1 |
NM_021458.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017384.mm.1 |
TC1400002252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017383.mm.1 |
NM_021458.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017383.mm.1 |
NM_021458.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017383.mm.1 |
TC1400002252.mm.1 |
MTA TC |