Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700030705.mm.1 |
NM_001252525.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030705.mm.1 |
NM_001252525.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030705.mm.1 |
NM_001252526.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030705.mm.1 |
NM_001252526.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030705.mm.1 |
NM_007755.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030705.mm.1 |
NM_007755.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030705.mm.1 |
TC0700003608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030706.mm.1 |
NM_001252525.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030706.mm.1 |
NM_001252525.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030706.mm.1 |
NM_001252526.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030706.mm.1 |
NM_001252526.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030706.mm.1 |
NM_007755.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030706.mm.1 |
NM_007755.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030706.mm.1 |
TC0700003608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030707.mm.1 |
NM_001252525.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030707.mm.1 |
NM_001252525.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030707.mm.1 |
NM_001252526.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030707.mm.1 |
NM_001252526.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030707.mm.1 |
NM_007755.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030707.mm.1 |
NM_007755.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030707.mm.1 |
TC0700003608.mm.1 |
MTA TC |