Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400016712.mm.1 |
NM_001164289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016712.mm.1 |
NM_001164289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016712.mm.1 |
TC1400002870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400020762.mm.1 |
NM_001164289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020762.mm.1 |
NM_001164289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020762.mm.1 |
TC1400002870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016714.mm.1 |
NM_001164289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016714.mm.1 |
NM_001164289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016714.mm.1 |
TC1400002870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016713.mm.1 |
NM_001164289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016713.mm.1 |
NM_001164289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016713.mm.1 |
TC1400002870.mm.1 |
MTA TC |