Assay Details
Gene Symbol: 4930434J06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046274.1 | 3 - 4 | 980 | 91 | ||
GenBank mRNA | AK015316.1 | - | 3 - 4 | 980 | 91 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | CA464707.1 | - | 353 | Chr. - - [-] on Build | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 4251 | Gm4251 | -590.6kb | Sense | Chr. 14 - 71251273 - 71271059 on NCBI Build GRCm38 | |
Downstream | fibronectin type III domain containing 3A | Fndc3a |
1700094E19Rik, D14Ertd453e, F730017H24Rik, Fndc3, sys |
639.1kb | Anti-sense | Chr. 14 - 72537948 - 72710686 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
