Assay Details
Gene Symbol: 4930434J06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046274.1 | 6 - 7 | 1350 | 75 | ||
GenBank mRNA | AK015316.1 | - | 6 - 7 | 1350 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 4251 | Gm4251 | -641.9kb | Sense | Chr. 14 - 71251273 - 71271059 on NCBI Build GRCm38 | |
Downstream | fibronectin type III domain containing 3A | Fndc3a |
1700094E19Rik, D14Ertd453e, F730017H24Rik, Fndc3, sys |
617.5kb | Anti-sense | Chr. 14 - 72537948 - 72710686 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2242510 | AK015316.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428719 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623057 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323857 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742865 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528187 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35927 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960065 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894288 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605711 | AF203444.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605711 | AK077478.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605711 | NM_019515.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605711 | XM_001475124.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597206 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318396 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427598 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392642 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672065 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546230 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740063 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593864 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655088 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 7679 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56460 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26907 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130303 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08157945 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08157904 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08158002 | - | - |
Whole Mouse Genome G4122A | A_52_P963032 | AK015316.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08158002 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08157904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709377 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709395 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709361 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709662 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709698 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709334 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709341 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709523 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709401 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709503 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709333 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709582 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709659 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709587 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11775130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11831271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709520 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709431 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709504 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11184652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709538 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709527 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709524 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709572 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709430 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709364 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11787380 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709536 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11714767 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709563 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709455 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709359 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709336 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709668 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709350 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709399 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709509 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11357826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11444727 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709591 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709387 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709571 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709584 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709366 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709373 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709656 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709528 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709561 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11310335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11627953 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709589 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709549 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709468 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709519 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709391 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709436 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709692 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709356 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709511 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709645 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709682 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709365 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11430423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709533 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709522 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11787364 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709384 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709541 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709460 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709461 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709570 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11310338 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709510 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709539 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709342 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709667 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11403537 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709428 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709690 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709512 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11444728 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11437932 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709701 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709368 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709379 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709703 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11440429 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11831282 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11444729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11687781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709403 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11701456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709695 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11775129 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709513 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709507 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709369 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11682582 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709516 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709393 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709531 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709381 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709467 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709447 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709351 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709592 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709542 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709358 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709529 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709343 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709551 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709425 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709664 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709526 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709547 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11430426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709530 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709347 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29709534 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1432030_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432030_at | - | - |
Mouse Genome 430 2.0 Array | 1432030_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400008174.mm.1 | NR_046274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008174.mm.1 | NR_046274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008174.mm.1 | TC1400001020.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400008173.mm.1 | NR_046274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008173.mm.1 | NR_046274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008173.mm.1 | TC1400001020.mm.1 | MTA TC |
Mouse Expression Set 430 | 1432030_PM_at | - | - |
Mouse Expression Set 430 | 1432030_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432030_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432030_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 519676 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 104610575 | - | - |
