Assay Details
Gene Symbol: Gm4251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046075.1 | 2 - 3 | 168 | 92 | ||
GenBank mRNA | AK076894.1 | - | 2 - 3 | 158 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503914 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588141 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265390 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554103 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512256 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37580 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005705 | AV269323.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005705 | AK076894.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08156518 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106671 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02880724 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706003 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760525 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706016 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706051 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706027 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11569107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11652959 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706031 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706078 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11443067 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706076 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706002 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705969 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11652972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11590228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706017 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11443047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706029 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706064 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11652965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706005 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11443066 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706066 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705973 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11279318 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706067 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760529 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11357388 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706015 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706023 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11387074 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705987 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705998 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11347833 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705990 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706069 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706021 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11192903 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11282126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706060 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11569108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760526 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706046 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11161720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29705989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706026 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29706035 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Mouse Genome 430 2.0 Array | 1458944_at | - | - |
Mouse Genome 430 2.0 Array | 1458944_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100043132_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100043132_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400008140.mm.1 | NR_046075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008140.mm.1 | NR_046075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008140.mm.1 | NR_046076.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008140.mm.1 | NR_046076.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008140.mm.1 | TC1400001013.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400008139.mm.1 | NR_046075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008139.mm.1 | NR_046075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008139.mm.1 | TC1400001013.mm.1 | MTA TC |
Mouse Expression Set 430 | 1458944_at | - | - |
Mouse Expression Set 430 | 1458944_PM_at | - | - |
Murine Genome U74v2 Set | 168795_at | - | - |
Murine Genome U74v2 Set | 168795_PM_at | - | - |
