Assay Details
| Gene Symbol: Slc28a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004184.3 | NP_001004184.2 | 7 - 8 | 811 | 55 | |
| GenBank mRNA | BC061230.1 | - | 7 - 8 | 811 | 30379959 | 55 |
More Information
Set Membership: |
|

Slc28a1
Genomic Map